Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXorf21 Polyclonal Antibody, Invitrogen™

Catalog No. FEPA5144179
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144179 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144179 Supplier Thermo Scientific Supplier No. PA5144179

Rabbit Polyclonal Antibody

CXorf21 Polyclonal antibody specifically detects CXorf21 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXorf21
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 3.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene CXorf21
Gene Accession No. Q9HAI6
Gene Alias 5430427O19Rik, chromosome X open reading frame 21, CXorf21, Protein CXorf21, uncharacterized protein CXorf21
Gene Symbols CXorf21
Host Species Rabbit
Immunogen Recombinant Human Uncharacterized protein CXorf21 protein (1-301AA)
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80231
Target Species Human
Content And Storage -20°C or -80°C if preferred
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.