Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXorf26 Monoclonal Antibody (OTI5A11), TrueMAB™, OriGene
SDP

Catalog No. 502541096
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1096 30 μL
50-167-4100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1096 Supplier OriGene Supplier No. TA503131S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CXorf26 Monoclonal antibody specifically detects CXorf26 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen CXorf26
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5A11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TUBB4A
Gene Accession No. P04350
Gene Alias 2610029G23Rik; CXorf26; MGC874; Pbdc1; polysaccharide biosynthesis domain containing 1; polysaccharide biosynthesis domain-containing protein 1; Protein PBDC1; UPF0368 protein Cxorf26; UPF0368 protein Cxorf26 homolog
Host Species Mouse
Immunogen Full length human recombinant protein of human TUBB4 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10382
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.