Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CXorf36 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551740
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551740 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551740 Supplier Invitrogen™ Supplier No. PA551740
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLPASSLSSL VPQVRTSYNF GRTFLGLDKC NACIGTSICK KFFKEEIRSD NWLASHLGLP PDSLLSYPAN YSDDSKIWRP VEIFRLVSKY QNEISDRRIC ASASAPKTCS IERVLRKTER FQKWLQAKRL TPDL Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 24%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXorf36
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CXorf36
Gene Accession No. Q9H7Y0
Gene Alias 4930578C19Rik; bA435K1.1; chromosome X open reading frame 36; CXorf36; deleted in autism-1 related protein; deleted in autism-related protein 1; DIA1R; DIPK2B; Divergent protein kinase domain 2B; EPQL1862; hCG1981635; PRO3743; UNQ1862/PRO3743; UPF0672 protein CXorf36
Gene Symbols CXorf36
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CXorf36. Recombinant protein control fragment (Product #RP-92129).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79742
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.