Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CXorf36 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546135
Description
Peptide sequence: DKQEGSQEAN RAGENKDIFS CLVSGCQAQL PSCESISEKQ SLVLVCQKLL Sequence homology: Horse: 79%; Human: 100%; Mouse: 79%.
The function of this protein remains unknown.
Specifications
CXorf36 | |
Polyclonal | |
Unconjugated | |
CXorf36 | |
4930578C19Rik; bA435K1.1; chromosome X open reading frame 36; CXorf36; deleted in autism-1 related protein; deleted in autism-related protein 1; DIA1R; DIPK2B; Divergent protein kinase domain 2B; EPQL1862; hCG1981635; PRO3743; UNQ1862/PRO3743; UPF0672 protein CXorf36 | |
Rabbit | |
Affinity chromatography | |
RUO | |
79742 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H7Y0 | |
CXorf36 | |
Synthetic peptide directed towards the C-terminal of human CXorf36 (aa 317-366). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction