Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CXorf38 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562139
Description
Immunogen sequence: DGCECEMGTY LSESQVNEIE MQLLKEKLQE IYLQAEEQEV LPEELSNRLE VVKEFLRNNE DLRNGLTEDM QKLDSLCLHQ KLDSQEPG Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 66%.
Specifications
| CXorf38 | |
| Polyclonal | |
| Unconjugated | |
| CXorf38 | |
| chromosome X open reading frame 38; CXorf38; Uncharacterized protein CXorf38 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 159013 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TB03 | |
| CXorf38 | |
| Recombinant protein corresponding to Human CXorf38. Recombinant protein control fragment (Product #RP-99967). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction