Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CXorf56 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558310
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558310 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558310 Supplier Invitrogen™ Supplier No. PA558310
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VSRSVVCSDT RDREEYDDGE KPLHVYYCLC GQMVLVLDCQ LEKLPMRPRD RSRVIDAAKH AHKFCNTEDE Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

While this gene is well-supported by transcript data, no functional information on its protein products is currently available. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXorf56
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CXorf56
Gene Accession No. Q9H5V9
Gene Alias chromosome X open reading frame 56; CXorf56; UPF0428 protein; UPF0428 protein CXorf56
Gene Symbols CXorf56
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CXorf56. Recombinant protein control fragment (Product #RP-95425).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63932
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.