Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CXorf57 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA535870
Description
Recommended positive controls: CXorf57-transfected 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The CXorf57 gene, located on the X chromosome, encodes a protein involved in various biological processes, although its precise functions remain under investigation. CXorf57 is expressed in a number of tissues, including the brain and reproductive organs, suggesting a role in neurodevelopment and fertility. Emerging research has highlighted its potential association with immune responses and cellular signaling pathways. Studies are focused on understanding the gene's involvement in disease mechanisms, particularly in the context of its expression patterns in different conditions. Further elucidation of CXorf57's functional roles could provide insights into its contributions to human health and disease.
Specifications
| CXorf57 | |
| Polyclonal | |
| Unconjugated | |
| CXorf57 | |
| chromosome X open reading frame 57; CXorf57; RADX; RP11-647M7.1; RPA-related and RAD51-antagonist, X-chromosome; RPA-related protein RADX; uncharacterized protein CXorf57 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55086 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.71 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q6NSI4 | |
| CXorf57 | |
| Recombinant protein encompassing a sequence within the center region of human CXorf57. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction