Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXorf61 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501735032 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5032 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5032 Supplier Proteintech Group Inc Supplier No. 257081AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CT83 is a protein coding gene.

Specifications

Antigen CXorf61
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CT83
Gene Accession No. Q5H943
Gene Alias Cancer/testis antigen 83, CT83, CXorf61, KK LC 1, KKLC1
Gene Symbols CT83
Host Species Rabbit
Immunogen CXorf61 Fusion Protein Ag22619
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 203413
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.