Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CXorf67 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544458
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544458 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544458 Supplier Invitrogen™ Supplier No. PA544458
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SSYASNSSSP SRSPGLSPSS PSPEFLGLRS ISTPSPESLR YALMPEFYAL Sequence homology: Dog: 91%; Human: 100%; Mouse: 83%.

Inhibits PRC2/EED-EZH2 complex function thereby causing down-regulation of H3K27me3. In the PRC2/EED-EZH2 complex, interacts and inhibits EZH2 methyltransferase activity. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXorf67
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CXorf67
Gene Accession No. Q86X51
Gene Alias chromosome X open reading frame 67; CXorf67; EZH inhibitory protein; EZHIP; uncharacterized protein CXorf67
Gene Symbols CXorf67
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human LOC340602.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340602
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.