Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXXC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142457
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142457 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142457 Supplier Thermo Scientific Supplier No. PA5142457
Add to Cart
Add to Cart
This item is not returnable. View return policy

CXXC4 Polyclonal Antibody

CXXC4 Polyclonal Antibody for IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen CXXC4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene CXXC4
Gene Accession No. Q9H2H0
Gene Alias 9330210J02Rik; C030003J12Rik; CXXC finger 4; CXXC finger protein 4; CXXC4; CXXC-type zinc finger protein 4; Dvl-binding protein IDAX (inhibition of the Dvl and Axin complex); Idax; inhibition of the Dvl and axin complex protein; inhibitor of the Dvl and Axin complex
Gene Symbols CXXC4
Host Species Goat
Immunogen Peptide with sequence C-GTSLERTPVPSAEA.
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80319
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.