Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CXXC5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566138
Description
Immunogen sequence: ASLLANGHDLA AAMAVDKSNP TSKHKSGAVA SLLSKAERAT ELAAEGQLTL QQFAQSTEML KRVVQEHLPL Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
CXXC5 is zinc finger protein closely related to the DVL1-binding protein CXXC4 (also known as Idax), and like CXXC4 is thought to modulate the Wnt signaling pathway during development. CXXC5 is upregulated in neural stem cells upon BMP4 stimulation, leading to decreased levels of the Wnt-signaling target AXIN2 and facilitated the response of the stem cells to Wnt3a. Recent reports suggest CXXC5 transcription is activated by the Wilms tumor 1 (WT1) transcription factor, and together are involved in the WNT/beta-catenin pathway and play a role in embryonic kidney development.
Specifications
CXXC5 | |
Polyclonal | |
Unconjugated | |
CXXC5 | |
4930415K17Rik; CF5; CXXC finger 5; CXXC finger 5 protein; CXXC finger protein 5; Cxxc5; CXXC-type zinc finger protein 5; HSPC195; Putative MAPK-activating protein PM08; putative NF-kappa-B-activating protein 102; Retinoid-inducible nuclear factor; RINF; TCCCIA00297; WID; WT1-induced Inhibitor of Dishevelled | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51523 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7LFL8 | |
CXXC5 | |
Recombinant Human CXXC5. Recombinant protein control fragment (Product #RP-105094). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction