Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CXXC5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104329
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104329 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104329 Supplier Invitrogen™ Supplier No. PA5104329
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CXXC5.

CXXC5 is zinc finger protein closely related to the DVL1-binding protein CXXC4 (also known as Idax), and like CXXC4 is thought to modulate the Wnt signaling pathway during development. CXXC5 is upregulated in neural stem cells upon BMP4 stimulation, leading to decreased levels of the Wnt-signaling target AXIN2 and facilitated the response of the stem cells to Wnt3a. Recent reports suggest CXXC5 transcription is activated by the Wilms tumor 1 (WT1) transcription factor, and together are involved in the WNT/beta-catenin pathway and play a role in embryonic kidney development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXXC5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CXXC5
Gene Accession No. Q5XIQ3, Q7LFL8, Q91WA4
Gene Alias 4930415K17Rik; CF5; CXXC finger 5; CXXC finger 5 protein; CXXC finger protein 5; Cxxc5; CXXC-type zinc finger protein 5; HSPC195; Putative MAPK-activating protein PM08; putative NF-kappa-B-activating protein 102; Retinoid-inducible nuclear factor; RINF; TCCCIA00297; WID; WT1-induced Inhibitor of Dishevelled
Gene Symbols CXXC5
Host Species Rabbit
Immunogen A synthesized peptide derived from human CXXC5(Accession Q7LFL8), corresponding to amino acid residues P46-M96.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 291670, 51523, 67393
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.