Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CY 09
GREENER_CHOICESDP

Catalog No. 643650 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-365-0 50 mg
64-361-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 64-365-0 Supplier Tocris Bioscience™ Supplier No. 6436/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Inflammasomes Inhibitors

CY 09 is a NLRP3 inhibitor (Kd = 500 nM); directly binds to ATP-binding motif of NLRP3 NACHT domain. Blocks NLRP3 inflammasome assembly and activation. Displays therapeutic effects on mouse models of cryopyrin-associated autoinflammatory syndrome (CAPS) and type 2 diabetes. Exhibits no activity for hERG (10 μM).

Specifications

Chemical Name or Material 4-[[4-Oxo-2-thioxo-3-[[3-(trifluoromethyl)phenyl]methyl]-5-thiazolidinylidene]methyl]benzoic acid
CAS 1073612-91-5
Quantity 50 mg
Molecular Formula C19H12F3NO3S2
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.