Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Cy™3B Mono-reactive NHS Ester Fluorescent Dyes

Catalog No. 09928187 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-928-187 25 mg
09-928-193 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-928-187 Supplier Cytiva Supplier No. PA63106
Add to Cart
Add to Cart
Request a Quote

Combines exceptional photostability with bright and intense signals. Cytiva Cy™3B NHS Ester Mono-Reactive CyDye™ provides targeted and specific labeling of amine residues on antibodies, peptides, or oligonucleotides in fluorescence polarization applications.

CyDye™ fluors are versatile fluorophores designed for a broad range of applications. CyDye™ fluors produce high sensitivity and highly reproducible performance at a biologically suitable pH range of 3 to 10. The use of multicolor CyDye™ flours enables multiplexing without crosstalk for high accuracy.

Cy™3B NHS Ester provides targeted and specific labeling of amine residues on antibodies, peptides, or oligonucleotides.

  • Readily dissolves in and produces a stable signal in aqueous buffers (no need for organic solvents)
  • High purity and thorough QC testing generate high levels of chromophore and reactive dye content
  • Narrow excitation and emission bands result in discrete signals from each fluor
  • Demonstrates larger polarization range than conventional fluors
  • Fluorescence lifetime of 2.8 ns
  • High fluorescence intensity
  • Low non-specific binding
TRUSTED_SUSTAINABILITY

Specifications

Formulation 100% Cy3B (NHS ester)
Format Solid
Quantity 25 mg
Product Type Cy3B NHS Ester
Molecular Weight (g/mol) 771
Purity >99%
Solubility Soluble in aqueous buffers
Label or Dye Cy3
Color Red
Emission 570
Excitation 559
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.