Learn More
Description
CyDye fluors are versatile fluorophores designed for a broad range of applications. CyDye fluors produce high sensitivity and highly reproducible performance at a biologically suitable pH range of 3 to 10. The use of multicolor CyDye flours enables multiplexing without crosstalk for high accuracy.
Cy3 NHS Ester provides targeted and specific labeling of amine residues on antibodies, peptides, or oligonucleotides.
- Readily dissolves in and produces a stable signal in aqueous buffers (no need for organic solvents)
- High purity and thorough QC testing generate high levels of chromophore and reactive dye content
- Narrow excitation and emission bands result in discrete signals from each fluor
- Demonstrates larger polarization range than conventional fluors
- Fluorescence lifetime of 2.8 ns
- High fluorescence intensity
- Low non-specific binding
Specifications
Specifications
| Format | Dried |
| For Use With (Application) | For labeling via Amine groups, CyDyes are suitable for use in applications such as Fluorescence resonance energy transfer (FRET), time resolved-Fluorescence resonance energy transfer (TR-FRET) and Fluorescence polarization (FP) |
| Quantity | 10 mg |
| Product Type | Cy3 NHS Ester |
| Kit Contents | Foil packs of pre-measured dried dye |
| pH Range | 3 to 10 |
| Solubility | Soluble in aqueous solutions |
| Content And Storage | 2°C to 8°C |
| Label or Dye | Cy3 |
| Color | Red |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.