Learn More
Description
- Excitation at 649 nm and emission at 671 nm.
- The fluorescent dye Cy5 is conjugated to the non-reducing end terminal GlcNAc residue.
- Heparan sulfate can be released by digestion with heparanase or heparinase II, heparinase III.
- Can be separated on 15%-17% SDS-PAGE and directly visualized.
Specifications
Specifications
| Quantity | 100 pmol |
| Product Line | R&D Systems |
| Target | Endorepellin/Perlecan/Heparan Sulfate Proteoglycan |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
