Cyanide Standard, 1mL == 1mg CN, 1000ppm CN, Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 25434

Packaging Amber Poly
Quantity 120mL

 View more versions of this product

Catalog No. 2543-4

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 151-50-8 (Potassium Cyanide),1310-73-2 (Sodium Hydroxide),7732-18-5 (Water, Deionized)
    Physical Form Liquid
    Chemical Name or Material Cyanide Standard

    Specifications

    Color Clear colorless
    Physical Form Liquid
    Quantity 120mL
    Concentration or Composition Notes 1000ppm CN
    Equivalence 1 mL = 1 mg CN
    CAS 151-50-8 (Potassium Cyanide),1310-73-2 (Sodium Hydroxide),7732-18-5 (Water, Deionized)
    Packaging Amber Poly
    Chemical Name or Material Cyanide Standard

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit