Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanine Sheep anti-Chemical, HRP, Polyclonal, Rockland™
SDP

Catalog No. 501992575
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2575 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-2575 Supplier Rockland Immunochemicals Supplier No. 600603209
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Sheep Serum, anti-Peroxidase and cyanine conjugated IgG. Reactivity was observed against Cy2, Cy3 and Cy5. Reactivity against other cyanine analog is unknown.

Cyanines are blue-green fluorescent dyes belonging to polymethine group, used in biotechnology for labeling and analysis. The spectrum for Cyanine ranges from IR (489/506) to UV (743/767). Cyanine dyes include Cy2, Cy3, Cy3B, Cy3.5, Cy5, Cy5.5 and Cy7. These dyes exhibit brighter and more stable fluorescence than traditional FITC, TRITC/Rhodamine dyes. Cy3 and Cy5 are most commonly used for 2 color detection in Flow Cytometry applications.

Specifications

Antigen Cyanine
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate HRP
Formulation 0.02M potassium phosphate with 0.15M NaCl, 10 mg/mL BSA and 0.01% gentamicin sulfate; pH 7.2
Gene Alias Cy dye; Cyanine dye
Host Species Sheep
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.