Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanogen bromide, Alfa Aesar™

Manufacturer:  Alfa Aesar 3273122

Quantity 100g

 View more versions of this product

Catalog No. AA3273122


 

Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Chemical Identifiers

506-68-3
105.93
UN1889
Cyanogen bromide
BrCN
MFCD00011597
14,2693

Specifications

2.015
61.4°C
100g
Crystalline
105.93
UN1889
Cyanogen bromide
Acrid
50° to 52°C
506-68-3
BrCN
MFCD00011597
14,2693

Safety and Handling

SDS & Safety Documents

GHS H Statement
H300-H310-H330;
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: CYANOGEN BROMIDE

RTECSNumber : GT2100000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit