Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanogen iodide, 98%, Thermo Scientific Chemicals

Catalog No. AC405960250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC405960250 25 g
AC405960050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC405960250 Supplier Thermo Scientific Chemicals Supplier No. 405960250

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-78-5
Molecular Formula CIN
InChI Key WPBXOELOQKLBDF-UHFFFAOYSA-N
Synonym cyanogen iodide, iodine cyanide, jodcyan, iodocyanide, iodine monocyanide, cyanogen monoiodide, iodine cyanide i cn, unii-81esv254uf, cyanic iodide, cyano iodide
PubChem CID 10478
IUPAC Name carbononitridic iodide
SMILES C(#N)I

Specifications

Melting Point 142°C to 149°C
Color Beige to Brown or Pink
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula ICN
Quantity 25 g
Molecular Weight (g/mol) 152.922
Formula Weight 152.92
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material Cyanogen iodide
Show More Show Less
Hazard Category GHS Signal Word: Danger

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.