Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(ethylene Glycol), Average M.W. 8000

Catalog No. AC418050010
Click to view available options
Quantity:
1 kg
3 kg
10 kg
Packaging:
Plastic Bottle
Plastic drum

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEG
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Color white flakes
Melting Point 55.0°C to 60.0°C
Linear Formula H(OCH2CH2)nOH
Merck Index 15, 7688
Quantity 1 kg
Flash Point 270°C
Hydroxyl Value 12 to 16mg KOH/g
Packaging Plastic Bottle
Viscosity 300-450 mPa.s (20°C 50% in water)
Water ≤0.5% (K.F.)
Chemical Name or Material Poly(ethylene glycol), Average M.W. 8000
Show More Show Less
EINECSNumber 500-038-2
RTECSNumber TQ4100000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.