Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanuric acid, 98%

Catalog No. AC181020010
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Plastic bottle

CAS: 108-80-5 | C3H3N3O3 | 129.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-80-5
Molecular Formula C3H3N3O3
Molecular Weight (g/mol) 129.08
MDL Number MFCD00082990
InChI Key ZFSLODLOARCGLH-UHFFFAOYSA-N
Synonym cyanuric acid, isocyanuric acid, 1,3,5-triazine-2,4,6-triol, trihydroxycyanidine, tricyanic acid, sym-triazinetriol, s-triazinetriol, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, pseudocyanuric acid, isocyanurate acid
PubChem CID 7956
ChEBI CHEBI:38028
IUPAC Name 1,3,5-triazinane-2,4,6-trione
SMILES O=C1NC(=O)NC(=O)N1

Specifications

Color White to Beige
Assay Percent Range 98%
Beilstein 26, 239
Merck Index 15, 2690
Formula Weight 129.07
Percent Purity ≥97.5%
Quantity 1 kg
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Cyanuric acid, 98%
Show More Show Less
Hazard Category HYGROSCOPIC
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-618-0
RTECSNumber XZ1800000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.