Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYB561 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103405
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103405 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103405 Supplier Invitrogen™ Supplier No. PA5103405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Cytochrome b561.

Secretory vesicle-specific electron transport protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYB561
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CYB561
Gene Accession No. P49447, Q60720
Gene Alias CD158F; CD158F1; Cyb561; CYB561A1; cytochrome b561; Cytochrome b-561; cytochrome b561 family, member A1; ferric-chelate reductase 2; FRRS2; KIR2DL5; Mcyt
Gene Symbols CYB561
Host Species Rabbit
Immunogen A synthesized peptide derived from human CYB561(Accession P49447), corresponding to amino acid residues H53-V103.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13056, 1534
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.