Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CYB561A3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5139806
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: ILLASSWKRP EPGILTDRQP LLHDGE.

Specifications
CYB561A3 | |
Polyclonal | |
Unconjugated | |
CYB561A3 | |
CYB561A3; CYBASC3; Cytochrome b ascorbate-dependent protein 3; cytochrome b, ascorbate dependent 3; cytochrome b561 family member A3; cytochrome b561 family, member A3; LCYTB; Lysosomal cytochrome b; PSEC0259 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
220002 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NBI2 | |
CYB561A3 | |
Recombinant protein corresponding to Human CYB561A3. Recombinant protein control fragment (Product #RP-100174). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction