Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYB561D1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599613
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599613 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599613 Supplier Invitrogen™ Supplier No. PA599613
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CYB561D1.

CYB561D1 gene ontology (GO) annotations include oxidation-reduction process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYB561D1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CYB561D1
Gene Accession No. A2AE42, Q8N8Q1
Gene Alias 1600010M23Rik; AI605489; AW457162; Cyb561d1; cytochrome b-561 domain containing 1; cytochrome b561 domain-containing protein 1; cytochrome b561 family member D1; cytochrome b561 family, member D1
Gene Symbols CYB561D1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CYB561D1(Accession Q8N8Q1), corresponding to amino acid residues A198-M229.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284613, 362017, 72023
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.