Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYB561D1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117900
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117900 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5117900 Supplier Invitrogen™ Supplier No. PA5117900
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CYB561D1 Polyclonal Antibody for Western Blot, IP

CYB561D1 gene ontology (GO) annotations include oxidation-reduction process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYB561D1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene CYB561D1
Gene Accession No. Q8N8Q1
Gene Alias 1600010M23Rik; AI605489; AW457162; Cyb561d1; cytochrome b-561 domain containing 1; cytochrome b561 domain-containing protein 1; cytochrome b561 family member D1; cytochrome b561 family, member D1
Gene Symbols CYB561D1
Host Species Rabbit
Immunogen E. coli-derived Human CYB561D1 fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284613
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.