Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CYB561D1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555488
Description
Immunogen sequence: MEDRSEGGRA RWVMPEIPAL WEADAGGSLE VFSPGTLYSW PWRSASAWLK PSYSSHLNTP CSSSAPEKHG SGSTGQGRP Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 30%.
CYB561D1 gene ontology (GO) annotations include oxidation-reduction process.
Specifications
CYB561D1 | |
Polyclonal | |
Unconjugated | |
CYB561D1 | |
1600010M23Rik; AI605489; AW457162; Cyb561d1; cytochrome b-561 domain containing 1; cytochrome b561 domain-containing protein 1; cytochrome b561 family member D1; cytochrome b561 family, member D1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
284613 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N8Q1 | |
CYB561D1 | |
Recombinant protein corresponding to Human CYB561D1. Recombinant protein control fragment (Product #RP-91297). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction