Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYB5B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552482
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552482 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552482 Supplier Invitrogen™ Supplier No. PA552482
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EVETSVTYYR LEEVAKRNSL KELWLVIHGR VYDVTRFLNE HPGGEEVLLE QAGVDASESF EDVGHSSDAR EMLKQYYIGD IHPSDLKPES GSKD Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 79%.

CYB5B gene ontology annotations related to this gene include enzyme activator activity; heme binding; metal ion binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYB5B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CYB5B
Gene Accession No. O43169
Gene Alias 1810044O22Rik; AU015618; Cyb5b; Cyb5m; CYB5-M; CYPB5M; cytochrome b5 outer mitochondrial membrane isoform; Cytochrome b5 type B; cytochrome b5 type B (outer mitochondrial membrane); cytochrome b5, outer mitochondrial membrane isoform; OMB5; outer mitochondrial membrane cytochrome b5; type 2 cyt-b5
Gene Symbols CYB5B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CYB5B. Recombinant protein control fragment (Product #RP-89712).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80777
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.