Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYB5B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597644
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597644 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597644 Supplier Invitrogen™ Supplier No. PA597644
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CYB5B Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

CYB5B gene ontology annotations related to this gene include enzyme activator activity; heme binding; metal ion binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYB5B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CYB5B
Gene Accession No. O43169, Q9CQX2
Gene Alias 1810044O22Rik; AU015618; Cyb5b; Cyb5m; CYB5-M; CYPB5M; cytochrome b5 outer mitochondrial membrane isoform; Cytochrome b5 type B; cytochrome b5 type B (outer mitochondrial membrane); cytochrome b5, outer mitochondrial membrane isoform; OMB5; outer mitochondrial membrane cytochrome b5; type 2 cyt-b5
Gene Symbols CYB5B
Host Species Rabbit
Immunogen Recombinant Human Cytochrome b5 type B protein (2-118AA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66427, 80777
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.