Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYB5R1 Antibody (OTI4B7), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271943B
Description
CYB5R1 Monoclonal specifically detects CYB5R1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
CYB5R1 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
CYB5R1 | |
Full length recombinant protein of human CYB5R1 (NP_057327) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI4B7 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
b5R.1, B5R1, CYB5BR1, cytochrome b5 reductase 1, EC 1.6.2.2, Humb5R2, NAD(P)H:quinone oxidoreductase type 3 polypeptide A2, NQO3A2polypeptide A2 | |
Mouse | |
Affinity Purified | |
metabolism | |
51706.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction