Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYB5R1 Monoclonal Antibody (OTI2D10), TrueMAB™, OriGene
SDP

Catalog No. 501675286
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-5286 100 μL
50-254-0200 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-5286 Supplier OriGene Supplier No. TA504639
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NADH-cytochrome b5 reductases are involved in desaturation and elongation of fatty acids, cholesterol biosynthesis, drug metabolism, and, in erythrocyte, methemoglobin reduction.

Specifications

Antigen CYB5R1
Applications Western Blot
Classification Monoclonal
Clone OTI2D10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CYB5R1
Gene Accession No. Q5EB81, Q9UHQ9
Gene Alias B5R.1, B5R1, B5R2, NQO3A2, humb5R2
Gene Symbols Cyb5r1
Host Species Mouse
Immunogen Full length human recombit protein of human CYB5R1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 304805, 51706, 606823
Target Species Canine, Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.