Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYB5R1 Monoclonal Antibody (OTI4B7)
GREENER_CHOICE

Catalog No. PIMA525929
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525929 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525929 Supplier Invitrogen™ Supplier No. MA525929
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CYB5R1 Monoclonal Antibody for ICC/IF

NADH-cytochrome b5 reductases are involved in desaturation and elongation of fatty acids, cholesterol biosynthesis, drug metabolism, and, in erythrocyte, methemoglobin reduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYB5R1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4B7
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CYB5R1
Gene Accession No. Q9UHQ9
Gene Alias 1500005G05Rik; b5R.1; B5R1; B5R2; C80155; Cyb5r1; cytochrome b5 reductase 1; Humb5R2; NAD(P)H:quinone oxidoreductase type 3 polypeptide A2; NAD(P)H:quinone oxidoreductase type 3, polypeptide A2; NAD(P)H:quinone oxidoreductase type 3, polypeptide A2; cytochrome b5 reductase 1; NADH-cytochrome b5 reductase 1; Nqo3a2; UNQ3049/PRO9865
Gene Symbols CYB5R1
Host Species Mouse
Immunogen Full length human recombinant protein of CYB5R1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51706
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.