Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYB5R1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550071
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550071 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550071 Supplier Invitrogen™ Supplier No. PA550071
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total CYB5R1 protein.

NADH-cytochrome b5 reductases are involved in desaturation and elongation of fatty acids, cholesterol biosynthesis, drug metabolism, and, in erythrocyte, methemoglobin reduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYB5R1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CYB5R1
Gene Accession No. Q5EB81, Q9DB73, Q9UHQ9
Gene Alias 1500005G05Rik; b5R.1; B5R1; B5R2; C80155; Cyb5r1; cytochrome b5 reductase 1; Humb5R2; NAD(P)H:quinone oxidoreductase type 3 polypeptide A2; NAD(P)H:quinone oxidoreductase type 3, polypeptide A2; NAD(P)H:quinone oxidoreductase type 3, polypeptide A2; cytochrome b5 reductase 1; NADH-cytochrome b5 reductase 1; Nqo3a2; UNQ3049/PRO9865
Gene Symbols CYB5R1
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human CYB5R1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 304805, 51706, 72017
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.