Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYB5R2 Antibody (OTI4B9), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271942R
Description
CYB5R2 Monoclonal specifically detects CYB5R2 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
CYB5R2 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CYB5R2 | |
Full length recombinant protein of human CYB5R2 (NP_057313) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot, Flow Cytometry | |
OTI4B9 | |
Western Blot, Flow Cytometry | |
B5R.2, cytochrome b5 reductase 2 | |
Mouse | |
Affinity Purified | |
Cardiovascular Biology | |
51700.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction