Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYB5R2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528038
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528038 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528038 Supplier Invitrogen™ Supplier No. PA528038
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: PC-3, U87-MG, SK-N-SH. Predicted reactivity: Mouse (85%), Rat (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CYB5R2 is an NADH-cytochrome b5 reductase which are involved in desaturation and elongation of fatty acids, cholesterol biosynthesis, drug metabolism, and, in erythrocyte, methemoglobin reduction. Responsible for NADH-dependent lucigenin chemiluminescence in spermatozoa by reducing both lucigenin and 2-[4-iodophenyl]-3-[4-nitrophenyl]-5-[2,4-disulfophenyl]-2H tetrazolium monosodium salt (WST-1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYB5R2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene CYB5R2
Gene Accession No. Q6BCY4
Gene Alias b5R.2; b5R2; CYB5R2; cytochrome b5 reductase 2; cytochrome b5 reductase b5R.2; D630003K02Rik; NADH-cytochrome b5 reductase 2; RGD1308421
Gene Symbols CYB5R2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CYB5R2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51700
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.