Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CYB5R2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558611
Description
Immunogen sequence: MTQYLENMKI GETIFFRGPR GRLFYHGPGN LGIRPDQTSE PKKTLADHLG MI Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 73%.
CYB5R2 is an NADH-cytochrome b5 reductase which are involved in desaturation and elongation of fatty acids, cholesterol biosynthesis, drug metabolism, and, in erythrocyte, methemoglobin reduction. Responsible for NADH-dependent lucigenin chemiluminescence in spermatozoa by reducing both lucigenin and 2-[4-iodophenyl]-3-[4-nitrophenyl]-5-[2,4-disulfophenyl]-2H tetrazolium monosodium salt (WST-1).
Specifications
CYB5R2 | |
Polyclonal | |
Unconjugated | |
CYB5R2 | |
b5R.2; b5R2; CYB5R2; cytochrome b5 reductase 2; cytochrome b5 reductase b5R.2; D630003K02Rik; NADH-cytochrome b5 reductase 2; RGD1308421 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51700 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6BCY4 | |
CYB5R2 | |
Recombinant protein corresponding to Human CYB5R2. Recombinant protein control fragment (Product #RP-97438). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction