Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYB5R2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569980
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569980 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569980 Supplier Invitrogen™ Supplier No. PA569980
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 92%; Dog: 92%; Guinea Pig: 92%; Horse: 85%; Human: 100%; Mouse: 92%; Rabbit: 100%; Rat: 100%.

CYB5R2 is an NADH-cytochrome b5 reductase which are involved in desaturation and elongation of fatty acids, cholesterol biosynthesis, drug metabolism, and, in erythrocyte, methemoglobin reduction. Responsible for NADH-dependent lucigenin chemiluminescence in spermatozoa by reducing both lucigenin and 2-[4-iodophenyl]-3-[4-nitrophenyl]-5-[2,4-disulfophenyl]-2H tetrazolium monosodium salt (WST-1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYB5R2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CYB5R2
Gene Accession No. Q6AY12
Gene Alias b5R.2; b5R2; CYB5R2; cytochrome b5 reductase 2; cytochrome b5 reductase b5R.2; D630003K02Rik; NADH-cytochrome b5 reductase 2; RGD1308421
Gene Symbols CYB5R2
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of rat CYB5R2 (aa 221-270).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 365345
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.