Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYB5R2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115074
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115074 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115074 Supplier Invitrogen™ Supplier No. PA5115074
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CYB5R2.

CYB5R2 is an NADH-cytochrome b5 reductase which are involved in desaturation and elongation of fatty acids, cholesterol biosynthesis, drug metabolism, and, in erythrocyte, methemoglobin reduction. Responsible for NADH-dependent lucigenin chemiluminescence in spermatozoa by reducing both lucigenin and 2-[4-iodophenyl]-3-[4-nitrophenyl]-5-[2,4-disulfophenyl]-2H tetrazolium monosodium salt (WST-1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYB5R2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene CYB5R2
Gene Accession No. Q3KNK3, Q6AY12, Q6BCY4
Gene Alias b5R.2; b5R2; CYB5R2; cytochrome b5 reductase 2; cytochrome b5 reductase b5R.2; D630003K02Rik; NADH-cytochrome b5 reductase 2; RGD1308421
Gene Symbols CYB5R2
Host Species Rabbit
Immunogen A synthesized peptide derived from human CYB5R2(Accession Q6BCY4), corresponding to amino acid residues L146-E196.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 320635, 365345, 51700
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.