Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYB5R2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549380
Description
Predicted to react with rat based on sequence homology.
NADH-cytochrome b5 reductases are involved in desaturation and elongation of fatty acids, cholesterol biosynthesis, drug metabolism, and, in erythrocyte, methemoglobin reduction. Responsible for NADH-dependent lucigenin chemiluminescence in spermatozoa by reducing both lucigenin and 2-[4-iodophenyl]-3-[4-nitrophenyl]-5-[2,4-disulfophenyl]-2H tetrazolium monosodium salt (WST-1).
Specifications
CYB5R2 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q6BCY4, Q3KNK3 | |
CYB5R2 | |
KLH conjugated synthetic peptide between 33-67 amino acids from the N-terminal region of human CYB5R2. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CYB5R2 | |
NADH-cytochrome b5 reductase 2, b5R2, CYB5R2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
320635, 51700 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction