Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYB5R3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117902
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117902 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5117902 Supplier Invitrogen™ Supplier No. PA5117902
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CYB5R3 Polyclonal Antibody for Western Blot, IHC (P), IP

CYB5R3 (NADH-cytochrome b5, reductase 3, diaphorase 1) is a 301 amino acid protein encoded by the human gene CYB5R3. CYB5R3 belongs to the flavoprotein pyridine nucleotide cytochrome reductase family and has two naturally occuring isoforms. Isoform 1 is anchored to the cytoplasmic side of the endoplasmic reticulum membrane and mitochondrion outer membrane, while isoform 2 is the soluble form found in erythrocytes. CYB5R3 is involved in the desaturation and elongation of fatty acids, cholesterol biosynthesis,drug metabolism and, in erythrocytes, methemoglobin reduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYB5R3
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene CYB5R3
Gene Accession No. P00387
Gene Alias 0610016L08Rik; 2500002N19Rik; B5R; C5BR; C85115; CB5R; Cyb5r3; cytochrome b5 reductase 3; DIA1; Dia-1; Diaphorase (NADH) (cytochrome b-5 reductase); diaphorase 1; diaphorase 1 (NADH); Diaphorase1; diaphorase-1; LOC100524254; LOW QUALITY PROTEIN: NADH-cytochrome b5 reductase 3-like; mutant NADH-cytochrome b5 reductase; Nadhcb5; NADH-cytochrome b5 reductase 3; NADH-cytochrome b5 reductase 3 membrane-bound form; NADH-cytochrome b5 reductase 3 soluble form; NADH-cytochrome b5 reductase 3-like; RNNADHCB5; WU:AL591952.1-001; WU:AL591952.1-002; WU:AL591952.1-003; WU:Cyb5r3
Gene Symbols CYB5R3
Host Species Rabbit
Immunogen E. coli-derived Human CYB5R3 fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1727
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.