Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CYB5R3 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142674
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
CYB5R3 (NADH-cytochrome b5, reductase 3, diaphorase 1) is a 301 amino acid protein encoded by the human gene CYB5R3. CYB5R3 belongs to the flavoprotein pyridine nucleotide cytochrome reductase family and has two naturally occuring isoforms. Isoform 1 is anchored to the cytoplasmic side of the endoplasmic reticulum membrane and mitochondrion outer membrane, while isoform 2 is the soluble form found in erythrocytes. CYB5R3 is involved in the desaturation and elongation of fatty acids, cholesterol biosynthesis,drug metabolism and, in erythrocytes, methemoglobin reduction.
Specifications
CYB5R3 | |
Polyclonal | |
Unconjugated | |
CYB5R3 | |
0610016L08Rik; 2500002N19Rik; B5R; C5BR; C85115; CB5R; Cyb5r3; cytochrome b5 reductase 3; DIA1; Dia-1; Diaphorase (NADH) (cytochrome b-5 reductase); diaphorase 1; diaphorase 1 (NADH); Diaphorase1; diaphorase-1; LOC100524254; LOW QUALITY PROTEIN: NADH-cytochrome b5 reductase 3-like; mutant NADH-cytochrome b5 reductase; Nadhcb5; NADH-cytochrome b5 reductase 3; NADH-cytochrome b5 reductase 3 membrane-bound form; NADH-cytochrome b5 reductase 3 soluble form; NADH-cytochrome b5 reductase 3-like; RNNADHCB5; WU:AL591952.1-001; WU:AL591952.1-002; WU:AL591952.1-003; WU:Cyb5r3 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
109754, 25035 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P20070, Q9DCN2 | |
CYB5R3 | |
Peptide with sequence CLPNLERVGHPKER (aa 284-297). | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction