Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYB5R4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553886
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553886 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553886 Supplier Invitrogen™ Supplier No. PA553886
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RKVKLMFFNK TEDDIIWRSQ LEKLAFKDKR LDVEFVLSAP ISEWNGKQGH ISPALLSEFL KRNLDKSKVL VCICGPVPFT EQGVRLLHDL NFSK Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 79%.

NCB5OR is a flavohemoprotein that contains functional domains found in both cytochrome b5 (CYB5A; MIM 250790) and CYB5 reductase (see MIM 250800).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYB5R4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cyb5r4
Gene Accession No. Q7L1T6
Gene Alias 2810034J18Rik; b5&b5R; b5/b5r; B5+B5R; b5b5r; C79736; cb5/cb5R; Cb5cb5r; Cyb5r4; cytochrome b5 reductase 4; cytochrome b-type NAD(P)H oxidoreductase; dJ676J13.1; flavohemoprotein b5/b5R; flavohemoprotein b5+b5R; hypothetical protein LOC553685; im:4727699; NADPH cytochrome B5 oxidoreductase; Ncb5or; N-terminal cytochrome b5 and cytochrome b5 oxidoreductase domain-containing protein; RP4-676J13.1; Unknown (protein for MGC:134229); zgc:112177
Gene Symbols Cyb5r4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CYB5R4. Recombinant protein control fragment (Product #RP-92394).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51167
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.