Learn More
Invitrogen™ CYB5R4 Polyclonal Antibody

Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.
Specifications
Specifications
| Antigen | CYB5R4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | Cyb5r4 |
| Gene Accession No. | Q3TDX8 |
| Gene Alias | 2810034J18Rik; b5&b5R; b5/b5r; B5+B5R; b5b5r; C79736; cb5/cb5R; Cb5cb5r; Cyb5r4; cytochrome b5 reductase 4; cytochrome b-type NAD(P)H oxidoreductase; dJ676J13.1; flavohemoprotein b5/b5R; flavohemoprotein b5+b5R; hypothetical protein LOC553685; im:4727699; NADPH cytochrome B5 oxidoreductase; Ncb5or; N-terminal cytochrome b5 and cytochrome b5 oxidoreductase domain-containing protein; RP4-676J13.1; Unknown (protein for MGC:134229); zgc:112177 |
| Gene Symbols | Cyb5r4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.