Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYB5R4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983203
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3203 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3203 Supplier Bioss Supplier No. BS14133R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NCB5OR is a flavohemoprotein that contains functional domains found in both cytochrome b5 (CYB5A; MIM 250790) and CYB5 reductase (see MIM 250800).

Specifications

Antigen CYB5R4
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Cyb5r4
Gene Accession No. Q7L1T6
Gene Alias 2810034J18Rik; b5&b5R; b5/b5r; b5+b5R; b5b5r; BOS_117; C79736; cb5/cb5R; Cb5cb5r; cyb5r4; Cytochrome b5 reductase 4; cytochrome b-type NAD(P)H oxidoreductase; dJ676J13.1; Flavohemoprotein b5/b5R; flavohemoprotein b5+b5R; hypothetical protein LOC553685; im:4727699; NADPH cytochrome B5 oxidoreductase; Ncb5or; N-terminal cytochrome b5 and cytochrome b5 oxidoreductase domain-containing protein; RP4-676J13.1; zgc:112177
Gene Symbols Cyb5r4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CYB5R4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51167
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.