Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYB5RL Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | CYB5RL |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17961920
![]() |
Novus Biologicals
NBP17961920UL |
20 μL |
Each for $206.00
|
|
|||||
NBP179619
![]() |
Novus Biologicals
NBP179619 |
100 μL |
Each for $487.50
|
|
|||||
Description
CYB5RL Polyclonal specifically detects CYB5RL in Human samples. It is validated for Western Blot.Specifications
CYB5RL | |
Polyclonal | |
Rabbit | |
NP_001026842 | |
606495 | |
Synthetic peptide directed towards the N terminal of human LOC606495. Peptide sequence KLNPETFVAFCIIAMDRLTKDTYRVRFALPGNSQLGLRPGQHLILRGIVD. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
cytochrome b5 reductase-like | |
CYB5RL | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title