Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CYB5RL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570199
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.
NADH-cytochrome b5 reductases are involved in desaturation and elongation of fatty acids, cholesterol biosynthesis, drug metabolism, and, in erythrocyte, methemoglobin reduction.
Specifications
CYB5RL | |
Polyclonal | |
Unconjugated | |
CYB5RL | |
2810410C14Rik; Cyb5rl; cytochrome b5 reductase like; cytochrome b5 reductase-like; mFLJ00377; NADH-cytochrome b5 reductase-like | |
Rabbit | |
Affinity Chromatography | |
RUO | |
606495 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6IPT4 | |
CYB5RL | |
synthetic peptide directed towards the N terminal region of human CYB5RL. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction