Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CYB5RL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560073
Description
Immunogen sequence: QLPWSYQEKT HFGHLGQDLI KELVSCCRRK PFALVCGSAE FTKDIARCLL CAGLTEDSYF LF Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 76%.
NADH-cytochrome b5 reductases are involved in desaturation and elongation of fatty acids, cholesterol biosynthesis, drug metabolism, and, in erythrocyte, methemoglobin reduction.
Specifications
CYB5RL | |
Polyclonal | |
Unconjugated | |
CYB5RL | |
2810410C14Rik; Cyb5rl; cytochrome b5 reductase like; cytochrome b5 reductase-like; mFLJ00377; NADH-cytochrome b5 reductase-like | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
606495 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6IPT4 | |
CYB5RL | |
Recombinant protein corresponding to Human CYB5RL. Recombinant protein control fragment (Product #RP-99849). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction