Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYBA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577937
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577937 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577937 Supplier Invitrogen™ Supplier No. PA577937
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: K562, THP-1, HL-60 Predicted Reactivity: Dog (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Cytochrome b is comprised of a light chain (alpha) and a heavy chain (beta). This gene encodes the light, alpha subunit which has been proposed as a primary component of the microbicidal oxidase system of phagocytes. Mutations in this gene are associated with autosomal recessive chronic granulomatous disease (CGD), that is characterized by the failure of activated phagocytes to generate superoxide, which is important for the microbicidal activity of these cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYBA
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene CYBA
Gene Accession No. P13498
Gene Alias b558; CYBA; cytochrome b light chain; cytochrome b(558) alpha chain; cytochrome b(558) alpha-subunit; cytochrome b, alpha polypeptide; cytochrome b-245 alpha chain; cytochrome b-245 light chain; cytochrome b-245, alpha polypeptide; cytochrome b558 alpha-subunit; cytochrome b558 subunit alpha; cytochrome beta-558; flavocytochrome b-558 alpha polypeptide; neutrophil cytochrome b 22 kDa polypeptide; nmf333; p22 phagocyte B-cytochrome; p22 phox; p22phox; p22-PHOX; superoxide-generating NADPH oxidase light chain subunit
Gene Symbols CYBA
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of Human p22phox. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1535
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.