Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CYBA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571642
Description
CYBA Polyclonal Antibody for Western Blot, ICC/IF
Cytochrome b is comprised of a light chain (alpha) and a heavy chain (beta). This gene encodes the light, alpha subunit which has been proposed as a primary component of the microbicidal oxidase system of phagocytes. Mutations in this gene are associated with autosomal recessive chronic granulomatous disease (CGD), that is characterized by the failure of activated phagocytes to generate superoxide, which is important for the microbicidal activity of these cells.
Specifications
CYBA | |
Polyclonal | |
Unconjugated | |
CYBA | |
b558; CYBA; cytochrome b light chain; cytochrome b(558) alpha chain; cytochrome b(558) alpha-subunit; cytochrome b, alpha polypeptide; cytochrome b-245 alpha chain; cytochrome b-245 light chain; cytochrome b-245, alpha polypeptide; cytochrome b558 alpha-subunit; cytochrome b558 subunit alpha; cytochrome beta-558; flavocytochrome b-558 alpha polypeptide; neutrophil cytochrome b 22 kDa polypeptide; nmf333; p22 phagocyte B-cytochrome; p22 phox; p22phox; p22-PHOX; superoxide-generating NADPH oxidase light chain subunit | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
1535 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P13498 | |
CYBA | |
KLH conjugated synthetic peptide between 117-145 amino acids from the C-terminal region of human CYBA. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction