Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYBB/NOX2 Antibody (NL7), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP141012MFV450
Description
CYBB/NOX2 Monoclonal antibody specifically detects CYBB/NOX2 in Human samples. It is validated for Flow Cytometry, Flow (Intracellular)Specifications
| CYBB/NOX2 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| NL7 | |
| Flow Cytometry, Flow (Intracellular) | |
| CGD, CGD91-phox, Cytochrome b(558) subunit beta, cytochrome b-245 heavy chain, cytochrome b-245, beta polypeptide, Cytochrome b558 subunit beta, EC 1.6.3, GP91-1, GP91PHOX, GP91-PHOX, Heme-binding membrane glycoprotein gp91phox, NADPH oxidase 2, Neutrophil cytochrome b 91 kDa polypeptide, NOX2chronic granulomatous disease, p22 phagocyte B-cytochrome, p91-PHOX, Superoxide-generating NADPH oxidase heavy chain subunit | |
| Cytochrome B245 solubilised in octyl-beta-glucopyranoside | |
| 0.1 mL | |
| Cancer, Endocrinology, Immunology | |
| 1536 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction